Merck
Serum metabolite signatures of type 2 diabetes mellitus complications.
Journal of proteome research (2014-09-24)
Tao Wu, Guoxiang Xie, Yan Ni, Tao Liu, Ming Yang, Huafeng Wei, Wei Jia, Guang Ji
ABSTRACT
A number of metabolic conditions, including hypoglycemia, high blood pressure (HBP), dyslipidemia, nerve damage and amputation, and vision problems, occur as a result of uncontrolled blood glucose levels over a prolonged period of time. The different components of diabetic complications are not independent but rather interdependent of each other, rendering the disease difficult to diagnose and control. The underlying pathogenesis of those components cannot be easily elucidated because of the heterogeneous, polygenic, and multifactorial nature of the disease. Metabonomics offers a snapshot of distinct biochemical variations that may reflect the unique metabolic phenotype under pathophysiological conditions. Here we report a mass-spectrometry-based metabonomic study designed to identify the distinct metabolic changes associated with several complications of type 2 diabetes mellitus (T2DM). The 292 patients recruited in the study were divided into five groups, including T2DM with HBP, T2DM with nonalcoholic fatty liver disease (NAFLD), T2DM with HBP and NAFLD, T2DM with HBP and coronary heart disease (CHD), and T2DM with HBP, NAFLD, and CHD. Serum differential metabolites were identified in each group of T2DM complication, mainly involving bile acid, fatty acid, amino acid, lipid, carbohydrate, steroids metabolism, and tricarboxylic acids cycle. These broad-spectrum metabolic changes emphasize the complex abnormalities present among these complications with elevated blood glucose levels, providing a novel strategy for stratifying patients with T2DM complications using blood-based metabolite markers.
Product Description
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dibutyl phthalate, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetohydroxamic acid, 98%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, Absolute - Acetone free
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
USP
Acetohydroxamic acid, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetonitrile, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.