Merck
  • Highly efficient Heck reactions of aryl bromides with n-butyl acrylate mediated by a palladium/phosphine-imidazolium salt system.
Highly efficient Heck reactions of aryl bromides with n-butyl acrylate mediated by a palladium/phosphine-imidazolium salt system.
Organic letters (2001-06-05)
C Yang, H M Lee, S P Nolan
ABSTRACT
[reaction: see text] A new phosphine-imidazolium salt, L.HBr (1, L = (1-ethylenediphenylphosphino-3-(mesityl))imidazol-2-ylidene), has been prepared. A combination of 0.5 mol % of Pd(dba)(2) and 0.5 mol % of L.HBr in the presence of 2 equiv of Cs(2)CO(3) as base has proven to be highly efficient in the Heck coupling reactions of aryl bromides (from electron-deficient to electron-rich aryl bromides) with n-butyl acrylate.
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor