Merck
Recent advances in the synthesis of ammonium-based rotaxanes.
Molecules (Basel, Switzerland) (2010-07-27)
Dominic Thibeault, Jean-François Morin
ABSTRACT
The number of synthetic methods enabling the preparation of ammonium-based rotaxanes has increased very rapidly in the past ten years. The challenge in the synthesis of rotaxanes results from the rather weak interactions between the ammonium-containing rod and the crown ether macrocycle in the pseudorotaxane structure that rely mostly on O*H hydrogen bonds. Indeed, no strong base or polar solvent that could break up H-bonding can be used during the formation of rotaxanes because the two components will separate as two distinct entities. Moreover, most of the reactions have to be performed at room temperature to favor the formation of pseudorotaxane in solution. These non-trivial prerequisites have been taken into account to develop efficient reaction conditions for the preparation of rotaxanes and those are described in detail along this review.
Sigma-Aldrich
N-Isopropyl-N-methyl-tert-butylamine, 98%