Merck
  • Identification of sn-glycerol-1-phosphate dehydrogenase activity from genomic information on a hyperthermophilic archaeon, Sulfolobus tokodaii strain 7.
Identification of sn-glycerol-1-phosphate dehydrogenase activity from genomic information on a hyperthermophilic archaeon, Sulfolobus tokodaii strain 7.
Bioscience, biotechnology, and biochemistry (2006-01-24)
Yosuke Koga, Mami Ohga, Masanari Tsujimura, Hiroyuki Morii, Yutaka Kawarabayasi
ABSTRACT
sn-Glycerol-1-phosphate dehydrogenase is responsible for the formation of sn-glycerol-1-phosphate, the backbone of membrane phospholipids of Archaea. This activity had never been detected in cell-free extract of Sulfolobus sp. Here we report the detection of this activity on the thermostable ST0344 protein of Sulfolobus tokodaii expressed in Escherichia coli, which was predicted from genomic information on S. tokodaii. This is another line of evidence for the general mechanism of sn-glycerol-1-phosphate formation by the enzyme.
Sigma-Aldrich
sn-Glycerol 1-phosphate lithium salt, ≥95.0% (TLC)