Merck
  • Synthesis and antimicrobial activity of monodisperse copper nanoparticles.
Synthesis and antimicrobial activity of monodisperse copper nanoparticles.
Colloids and surfaces. B, Biointerfaces (2015-02-28)
Tomasz Kruk, Krzysztof Szczepanowicz, Joanna Stefańska, Robert P Socha, Piotr Warszyński
ABSTRACT
Metallic monodisperse copper nanoparticles at a relatively high concentration (300 ppm CuNPs) have been synthesized by the reduction of copper salt with hydrazine in the aqueous SDS solution. The average particles size and the distribution size were characterized by Dynamic Light Scattering (DLS), Nanosight-Nanoparticle Tracking Analysis (NTA). The morphology and structure of nanoparticles were investigated using Scanning Electron Microscopy (SEM). The chemical composition of the copper nanoparticles was determined by X-ray Photoelectron Spectroscopy (XPS). Monodisperse copper nanoparticles with average diameter 50 nm were received. UV/vis absorption spectra confirmed the formation of the nanoparticles with the characteristic peak 550 nm. The antimicrobial studies showed that the copper nanoparticles had high activity against Gram-positive bacteria, standard and clinical strains, including methicillin-resistant Staphylococcus aureus, comparable to silver nanoparticles and some antibiotics. They also exhibited antifungal activity against Candida species.
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF