Merck
  • Walphos versus Biferrocene-Based Walphos Analogues in the Asymmetric Hydrogenation of Alkenes and Ketones.
Walphos versus Biferrocene-Based Walphos Analogues in the Asymmetric Hydrogenation of Alkenes and Ketones.
Organometallics (2014-05-06)
Afrooz Zirakzadeh, Manuela A Groß, Yaping Wang, Kurt Mereiter, Walter Weissensteiner
ABSTRACT
Two representative Walphos analogues with an achiral 2,2″-biferrocenediyl backbone were synthesized. These diphosphine ligands were tested in the rhodium-catalyzed asymmetric hydrogenation of several alkenes and in the ruthenium-catalyzed hydrogenation of two ketones. The results were compared with those previously obtained on using biferrocene ligands with a