MilliporeSigma
  • Cloning of a novel family of mammalian GTP-binding proteins (RagA, RagBs, RagB1) with remote similarity to the Ras-related GTPases.
Cloning of a novel family of mammalian GTP-binding proteins (RagA, RagBs, RagB1) with remote similarity to the Ras-related GTPases.
The Journal of biological chemistry (1995-12-01)
A Schürmann, A Brauers, S Massmann, W Becker, H G Joost
ABSTRACT
cDNA clones of two novel Ras-related GTP-binding proteins (RagA and RagB) were isolated from rat and human cDNA libraries. Their deduced amino acid sequences comprise four of the six known conserved GTP-binding motifs (PM1, -2, -3, G1), the remaining two (G2, G3) being strikingly different from those of the Ras family, and an unusually large C-terminal domain (100 amino acids) presumably unrelated to GTP binding. RagA and RagB differ by seven conservative amino acid substitutions (98% identity), and by 33 additional residues at the N terminus of RagB. In addition, two isoforms of RagB (RagBs and RagB1) were found that differed only by an insertion of 28 codons between the GTP-binding motifs PM2 and PM3, apparently generated by alternative mRNA splicing. Polymerase chain reaction amplification with specific primers indicated that both long and short form of RagB transcripts were present in adrenal gland, thymus, spleen, and kidney, whereas in brain, only the long form RagB1 was detected. A long splicing variant of RagA was not detected. Recombinant glutathione S-transferase (GST) fusion proteins of RagA and RagBs bound large amounts of radiolabeled GTP gamma S in a specific and saturable manner. In contrast, GTP gamma S binding of GST-RagB1 hardly exceeded that of recombinant GST. GTP gamma S bound to recombinant RagA, and RagBs was rapidly exchangeable for GTP, whereas no intrinsic GTPase activity was detected. A multiple sequence alignment indicated that RagA and RagB cannot be assigned to any of the known subfamilies of Ras-related GTPases but exhibit a 52% identity with a yeast protein (Gtr1) presumably involved in phosphate transport and/or cell growth. It is suggested that RagA and RagB are the mammalian homologues of Gtr1 and that they represent a novel subfamily of Ras-homologous GTP binding proteins.