Merck
  • Monocarboxylate transporters 1-4 in NSCLC: MCT1 is an independent prognostic marker for survival.
Monocarboxylate transporters 1-4 in NSCLC: MCT1 is an independent prognostic marker for survival.
PloS one (2014-09-17)
Marte Eilertsen, Sigve Andersen, Samer Al-Saad, Yury Kiselev, Tom Donnem, Helge Stenvold, Ingvild Pettersen, Khalid Al-Shibli, Elin Richardsen, Lill-Tove Busund, Roy M Bremnes
ABSTRACT
Monocarboxylate transporters (MCTs) 1-4 are lactate transporters crucial for cancers cells adaption to upregulated glycolysis. Herein, we aimed to explore their prognostic impact on disease-specific survival (DSS) in both cancer and tumor stromal cells in NSCLC. Tissue micro arrays (TMAs) were constructed, representing both cancer and stromal tumor tissue from 335 unselected patients diagnosed with stage I-IIIA NSCLC. Immunohistochemistry was used to evaluate the expression of MCT1-4. In univariate analyses; ↓ MCT1 (P = 0.021) and ↑ MCT4 (P = 0.027) expression in cancer cells, and ↑ MCT1 (P = 0.003), ↓ MCT2 (P = 0.006), ↓ MCT3 (P = 0.020) expression in stromal cells correlated significantly with a poor DSS. In multivariate analyses; ↓ MCT1 expression in cancer cells (HR: 1.9, CI 95%: 1.3-2.8, P = 0.001), ↓ MCT2 (HR: 2.4, CI 95%: 1.5-3.9, P<0.001), ↓ MCT3 (HR: 1.9, CI 95%: 1.1-3.5, P = 0.031) and ↑ MCT1 expression in stromal cells (HR: 1.7, CI 95%: 1.1-2.7, P = 0.016) were significant independent poor prognostic markers for DSS. We provide novel information of MCT1 as a candidate marker for prognostic stratification in NSCLC. Interestingly, MCT1 shows diverging, independent prognostic impact in the cancer cell and stromal cell compartments.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
Anti-SLC16A1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
SAFC
BIS-TRIS