Merck
  • Phosphorylation and ubiquitylation are opposing processes that regulate endocytosis of the water channel aquaporin-2.
Phosphorylation and ubiquitylation are opposing processes that regulate endocytosis of the water channel aquaporin-2.
Journal of cell science (2014-05-31)
Hanne B Moeller, Takwa Shaiman Aroankins, Joachim Slengerik-Hansen, Trairak Pisitkun, Robert A Fenton
ABSTRACT
The post-translational modifications (PTMs) phosphorylation and ubiquitylation regulate plasma membrane protein function. Here, we examine the interplay between phosphorylation and ubiquitylation of the membrane protein aquaporin-2 (AQP2) and demonstrate that phosphorylation can override the previously suggested dominant endocytic signal of K63-linked polyubiquitylation. In polarized epithelial cells, although S256 is an important phosphorylation site for AQP2 membrane localization, the rate of AQP2 endocytosis was reduced by prolonging phosphorylation specifically at S269. Despite their close proximity, AQP2 phosphorylation at S269 and ubiquitylation at K270 can occur in parallel, with increased S269 phosphorylation and decreased AQP2 endocytosis occurring when K270 polyubiquitylation levels are maximal. In vivo studies support this data, with maximal levels of AQP2 ubiquitylation occurring in parallel to maximal S269 phosphorylation and enhanced AQP2 plasma membrane localization. In conclusion, we demonstrate for the first time that although K63-linked polyubiquitylation marks AQP2 for endocytosis, site-specific phosphorylation can counteract polyubiquitylation to determine its final localization. Similar mechanisms might exist for other plasma membrane proteins.
Product Description
Supelco
Deltamethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Supelco
Deltamethrin, PESTANAL®, analytical standard
Supelco
Forskolin, analytical standard
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution