Merck
  • Manipulating gene expression and signaling activity in cultured mouse limb bud cells.
Manipulating gene expression and signaling activity in cultured mouse limb bud cells.
Developmental dynamics : an official publication of the American Association of Anatomists (2014-03-19)
Jordan P Lewandowski, Taylor A Pursell, Adam H Rabinowitz, Steven A Vokes
ABSTRACT
The vertebrate limb bud is a well-established system for studying the mechanisms driving growth and patterning of an embryonic tissue. However, approaches for manipulating gene expression are currently limited to time-consuming methods. Culturing primary limb bud cells could potentially be used as a quicker assay. However, limb cells in culture quickly differentiate into cartilage under normal conditions, and approaches delivering DNA and siRNA into primary limb cells in culture are limited. These technical limitations have restricted the utility of limb buds for investigating problems that require higher-throughput approaches. In this report, we describe adaptations to a method for culturing primary limb bud cells in a pre-chondrogenic state, and generate a population of mouse primary limb cells that are responsive to Hedgehog (Hh) signaling. Hh-stimulated cells upregulate Hh target genes as well as an exogenous Hh-responsive reporter. We then describe a method for highly efficient delivery of plasmids and siRNAs into cultured primary limb bud cells in a 96-well format. Cultures of primary limb bud cells are amenable to gene manipulation under conditions that maintain the limb cells in an Hh-responsive, undifferentiated state. This approach provides a medium-throughput system to manipulate gene expression, and test DNA regulatory elements.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdk6
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK6
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Purmorphamine, A cell-permeable activator of Hedgehog signaling that induces osteoblast differentiation of multipotent mesenchymal progenitor cells C3H10T1/2 (EC₅₀ = 1 µM).
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
MISSION® siRNA Universal Negative Control #1
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)