Merck
  • Assessment of diagnostic and therapeutic approaches of Helicobacter pylori-associated iron deficiency and anemia in children with dyspeptic symptoms.
Assessment of diagnostic and therapeutic approaches of Helicobacter pylori-associated iron deficiency and anemia in children with dyspeptic symptoms.
Journal of the Egyptian Society of Parasitology (2015-02-04)
Mohiee El-Deen El-Aziz Awad, Saleh Mohamed Amin, Saied Mohamed Abdou
ABSTRACT
This study assessed the diagnostic approaches of Helicobacter pylori (IP)-associated iron deficiency (ID) and anemia (IDA) in children with dyspeptic symptoms and evaluated the effect of simultaneous anti-H. pylori (anti-HIP) therapy and oral iron in comparison with each of anti? HP therapy and oral iron therapy alone, on iron status as assessed by serum soluble transferrin receptor (sTfR) level. Two hundreds children with dyspeptic symptoms were subjected to clinical evaluation, stool examination, CBC, biochemical assays for serum iron parameters and measurements of serum IgG antibodies to HP and serum sTfR level by ELISA. Sixty children were found to have HP. associated ID or IDA and were randomly divided into 3 groups (20 children each). GA received 2-week anti-HP therapy plus 90-day oral iron, and GB received 2-week anti-HP therapy alone whereas group C received 90-day oral iron alone. Re-evaluation of the 3 groups was performed after 3 months of treatment initiation by repeat CBC and serum sTfR level. Children (45%) were HP-seropositive. The mean values of serum sTfR were significantly higher in HP-positive group and in HP-positive children with IDA than in HP-negative group and in HP-negative children with IDA although no significant differences were noted in hematologic variables and iron parameters between the corresponding groups and children. As regard treatment groups, there were significant improvements in the mean values of indices of IDA status (HIb, MCH, MCV, sTfR) and ID status (sTtRi) at 3 months of treatment initiation compared with their baseline values after. anti-HP triple therapy either with oral iron or without oral iron whereas the control children who were treated with oral iron alone showed insignificant changes despite oral iron administration. The improvements in these parameters were significantly greater in groups of children who received anti-HP therapy either combined with iron or alone, where compared with those who did not receive anti-HP therapy. Their magnitudes were significantly higher among children receiving anti-HP therapy combined with oral iron when compared with that receiving anti-HP therapy alone.
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Iron, tube, 100mm, outside diameter 10.0mm, inside diameter 7mm, wall thickness 1.5mm, annealed, 99.5%
Iron, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.998%
Iron, tube, 500mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.5%
Iron, tube, 200mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, wire reel, 100m, diameter 0.065mm, hard, 99.5%
Iron, foil, 300x300mm, thickness 0.15mm, hard, 99.5%
Iron, tube, 500mm, outside diameter 6.35mm, inside diameter 5.35mm, wall thickness 0.5mm, as drawn, 99.8%
Iron, tube, 1000mm, outside diameter 3.2mm, inside diameter 2.2mm, wall thickness 0.5mm, as drawn, 99.8%
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, tube, 200mm, outside diameter 6.35mm, inside diameter 5.35mm, wall thickness 0.5mm, as drawn, 99.8%
Iron, foil, not light tested, 50x50mm, thickness 0.001mm, 99.85%
Iron, foil, 2m coil, thickness 0.125mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.025mm, hard, 99.5%
Iron, foil, 2m coil, thickness 0.15mm, hard, 99.5%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 2m coil, thickness 0.38mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.38mm, hard, 99.5%
Iron, foil, 2m coil, thickness 0.038mm, hard, 99.5%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, foil, not light tested, 25x25mm, thickness 0.002mm, 99.85%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, not light tested, 300x300mm, thickness 0.05mm, hard, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%