Merck
  • Immune response profile elicited by the model antigen ovalbumin expressed in fusion with the bacterial OprI lipoprotein.
Immune response profile elicited by the model antigen ovalbumin expressed in fusion with the bacterial OprI lipoprotein.
Molecular immunology (2014-12-04)
Afonso P Basto, Marina Badenes, Sílvia C P Almeida, Carlos Martins, António Duarte, Dulce M Santos, Alexandre Leitão
ABSTRACT
The use of immunogenic formulations targeting pattern recognition receptors towards modulation of immune responses is a promising strategy to develop better vaccines against infectious and malignant diseases. Molecules targeting TLR2 offer interesting properties that are relevant for vaccine development, including the possibility to covalently attach the lipidic ligands to the antigens. However, the type of immune response elicited by these formulations is still controversial. In this work, we used the model antigen ovalbumin (OVA) expressed in fusion with the bacterial lipoprotein OprI in order to characterize the immunomodulatory properties of this TLR ligand. Murine bone marrow-derived dendritic cells stimulated with OprI-OVA fusion lipoprotein produced high levels of the pro-inflammatory cytokines TNF-α and IL-6 and also IL-10, IL-12(p70) and IL-27, while TGF-β and IL-23 were not detected. Using OT-II and OT-I mice, an enhancement of MHC class II and class I antigen presentation was observed for the OVA antigen in fusion with OprI. Mice immunized by intraperitoneal route with this fusion lipoprotein in prime-boost protocols developed strong specific antibody responses including IgG1, IgG2c, IgG2b, IgG3 and IgE. These results, together with data obtained by restimulation of splenocytes from the immunized mice, point to an immune response profile that does not correspond to a strict Th1 or Th2 polarization. Finally, in a challenge experiment using a melanoma syngeneic mouse model (B16-OVA), prophylactic inoculation with OprI fused with the unrelated antigen eGFP increased the tumor growth, while the fusion with the tumor-associated antigen OVA delayed the tumor growth and increased mice survival.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Imidazole, European Pharmacopoeia (EP) Reference Standard
SAFC
HEPES
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Albumin from chicken egg white, powder, 62-88% (agarose gel electrophoresis)
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)