Merck
  • Oxidation of diclofenac by potassium ferrate (VI): reaction kinetics and toxicity evaluation.
Oxidation of diclofenac by potassium ferrate (VI): reaction kinetics and toxicity evaluation.
The Science of the total environment (2014-12-03)
Yingling Wang, Haijin Liu, Guoguang Liu, Youhai Xie, Shuyan Gao
ABSTRACT
The reaction kinetics and toxicity of diclofenac (DCF) oxidation by ferrate (VI) under simulated water disinfection conditions were investigated. Experimental results indicated that the reaction between DCF and Fe(VI) followed first-order kinetics with respect to each reactant. Furthermore, the effects of pH and temperature on DCF oxidation by Fe(VI) were elucidated using a systematic examination. The apparent second-order rate constants (kapp) increased significantly from 2.54 to 11.6M(-1)s(-1), as the pH of the solution decreased from 11.0 to 7.0, and the acid-base equilibriums of Fe(VI) and DCF were proposed to explain the pH dependence of kapp. The acute toxicity of DCF solution during Fe(VI) oxidation was evaluated using a Microtox bioassay. Overall, the DCF degradation process resulted in a rapid increase of the inhibition rate of luminescent bacteria. These toxicity tests suggest that the formation of enhanced toxic intermediates during the Fe(VI) disinfection process may pose potential health risk to consumers.
Product Description
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Supelco
Diclofenac sodium salt, analytical standard
Diclofenac for system suitability, European Pharmacopoeia (EP) Reference Standard
Diclofenac potassium, European Pharmacopoeia (EP) Reference Standard
Diclofenac sodium, European Pharmacopoeia (EP) Reference Standard
USP
Diclofenac sodium, United States Pharmacopeia (USP) Reference Standard