Sigma-Aldrich
Oxalic acid, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Iron, IRMM®, certified reference material, 0.5 mm wire
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Iron(III) citrate tribasic monohydrate, 18-20% Fe basis (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ferric citrate, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Iron(III) citrate, technical grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal (751-770) Rabbit pAb, liquid, Calbiochem®
SKU
Pack Size
Availability
Price
Quantity
Iron, foil, not light tested, 50x50mm, thickness 0.001mm, 99.85%
SKU
Pack Size
Availability
Price
Quantity
Iron, foil, not light tested, 100x100mm, thickness 0.006mm, 99.85%
SKU
Pack Size
Availability
Price
Quantity
Iron, foil, not light tested, 150x150mm, thickness 0.008mm, 99.85%
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
Iron, wire reel, 1m, diameter 0.025mm, as drawn, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Iron, wire reel, 1m, diameter 1.0mm, as drawn, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Iron, tube, 500mm, outside diameter 12.0mm, inside diameter 10mm, wall thickness 1.0mm, annealed, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Iron, wire reel, 20m, diameter 0.25mm, hard, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Iron, foil, 10mm disks, thickness 0.075mm, as rolled, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Iron, foil, 1m coil, thickness 0.5mm, hard, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Iron, foil, 10mm disks, thickness 0.075mm, hard, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Iron, foil, 25mm disks, thickness 0.003mm, 99.85%