Merck
  • Significance of αThr-349 in the catalytic sites of Escherichia coli ATP synthase.
Significance of αThr-349 in the catalytic sites of Escherichia coli ATP synthase.
Biochemistry (2014-11-07)
Zulfiqar Ahmad, Mumeenat Winjobi, M Anaul Kabir
ABSTRACT
This paper describes the role of α-subunit VISIT-DG sequence residue αThr-349 in the catalytic sites of Escherichia coli F1Fo ATP synthase. X-ray structures show the highly conserved αThr-349 in the proximity (2.68 Å) of the conserved phosphate binding residue βR182 in the phosphate binding subdomain. αT349A, -D, -Q, and -R mutations caused 90-100-fold losses of oxidative phosphorylation and reduced ATPase activity of F1Fo in membranes. Double mutation αT349R/βR182A was able to partially compensate for the absence of known phosphate binding residue βR182. Azide, fluoroaluminate, and fluoroscandium caused insignificant inhibition of αT349A, -D, and -Q mutants, slight inhibition of the αT349R mutant, partial inhibition of the αT349R/βR182A double mutant, and complete inhibition of the wild type. Whereas NBD-Cl (7-chloro-4-nitrobenzo-2-oxa-1,3-diazole) inhibited wild-type ATPase and its αT349A, -D, -R, and -Q mutants essentially completely, βR182A ATPase and double mutant αT349A/βR182A were inhibited partially. Inhibition characteristics supported the conclusion that NBD-Cl reacts in βE (empty) catalytic sites, as shown previously by X-ray structure analysis. Phosphate protected against NBD-Cl inhibition in the wild type, αT349R, and double mutant αT349R/βR182A but not in αT349A, αT349D, or αT349Q. The results demonstrate that αThr-349 is a supplementary residue involved in phosphate binding and transition state stabilization in ATP synthase catalytic sites through its interaction with βR182.
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
SAFC
L-Threonine
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Aluminum, wire reel, 1000m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Aluminum, honeycomb, 600x600mm, thickness 5mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, foam, 150x150mm, thickness 30mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Aluminum, honeycomb, 600x600mm, thickness 20mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, honeycomb, 300x300mm, thickness 10mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, honeycomb, 300x300mm, thickness 52.3mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin glass fiber 0.75mm, 99.9%
Aluminum, wire reel, 50m, diameter 1.0mm, as drawn, 99.95+%
Aluminum, wire reel, 50m, diameter 1.0mm, as drawn, 99.999%
Aluminum, wire reel, 10m, diameter 0.125mm, as drawn, 99.5%
Aluminum, wire reel, 500m, diameter 0.25mm, as drawn, 99.5%
Aluminum, wire reel, 20m, diameter 0.063mm, annealed, 99.99%
Aluminum, wire reel, 1m, diameter 0.125mm, as drawn, 99.99+%
Aluminum, wire reel, 5m, diameter 1.5mm, as drawn, 99.999%
Aluminum, wire reel, 50m, diameter 0.063mm, annealed, 99.99%