Merck
  • Differential effects of exercise on brain opioid receptor binding and activation in rats.
Differential effects of exercise on brain opioid receptor binding and activation in rats.
Journal of neurochemistry (2014-10-21)
Ricardo Mario Arida, Sérgio Gomes da Silva, Alexandre Aparecido de Almeida, Esper Abrão Cavalheiro, Cecilia Zavala-Tecuapetla, Serge Brand, Luisa Rocha
ABSTRACT
Physical exercise stimulates the release of endogenous opioid peptides supposed to be responsible for changes in mood, anxiety, and performance. Exercise alters sensitivity to these effects that modify the efficacy at the opioid receptor. Although there is evidence that relates exercise to neuropeptide expression in the brain, the effects of exercise on opioid receptor binding and signal transduction mechanisms downstream of these receptors have not been explored. Here, we characterized the binding and G protein activation of mu opioid receptor, kappa opioid receptor or delta opioid receptor in several brain regions following acute (7 days) and chronic (30 days) exercise. As regards short- (acute) or long-term effects (chronic) of exercise, overall, higher opioid receptor binding was observed in acute-exercise animals and the opposite was found in the chronic-exercise animals. The binding of [(35) S]GTPγS under basal conditions (absence of agonists) was elevated in sensorimotor cortex and hippocampus, an effect more evident after chronic exercise. Divergence of findings was observed for mu opioid receptor, kappa opioid receptor, and delta opioid receptor receptor activation in our study. Our results support existing evidence of opioid receptor binding and G protein activation occurring differentially in brain regions in response to diverse exercise stimuli. We characterized the binding and G protein activation of mu, kappa, and delta opioid receptors in several brain regions following acute (7 days) and chronic (30 days) exercise. Higher opioid receptor binding was observed in the acute exercise animal group and opposite findings in the chronic exercise group. Higher G protein activation under basal conditions was noted in rats submitted to chronic exercise, as visible in the depicted pseudo-color autoradiograms.
Sigma-Aldrich
U-69593, solid
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
Naloxone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Naloxone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®