Merck
  • Long-term effectiveness of an ad hoc tailored titanium implant as a spacer for microvascular decompression in the treatment of trigeminal neuralgia caused by megadolichoectatic basilar artery anomaly: 9-year follow-up.
Long-term effectiveness of an ad hoc tailored titanium implant as a spacer for microvascular decompression in the treatment of trigeminal neuralgia caused by megadolichoectatic basilar artery anomaly: 9-year follow-up.
Journal of neurosurgery (2014-09-27)
Péter Banczerowski, Gábor Czigléczki, István Nyáry
ABSTRACT
An enlarged, elongated, ectatic, and sclerotic aberration of the vertebrobasilar system is known as a megadolichoectatic basilar artery (BA) anomaly. The anomaly is often involved in the pathological process of trigeminal neuralgia by compressing and distorting the trigeminal nerve. First-line medical treatment includes drug therapy, but a second-line surgical procedure could be effective in medication-resistant cases. The authors report the case of a 65-year-old man with a 12-year history of progressing trigeminal neuralgia who underwent microvascular decompression after the first-line drug treatment had failed. This case is unique because an in situ tailored titanium microplate was used as a spacer to alleviate compression by the BA on the trigeminal nerve. The titanium implant provided durable and sufficient retraction for the sclerotic arterial complex when the trigeminal nerve was placed in the tunnel of the implant. The 9-year follow-up examination proves the safety and long-term efficacy of titanium implants in the treatment of trigeminal neuralgia caused by a megadolichoectatic BA anomaly. The method applied in this case was not intended to be and certainly is not an alternative to routine microvascular decompression-this surgical solution may be reserved for some extreme cases.
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Titanium, microfoil, disks, 10mm, thinness 0.1μm, specific density 42.8μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, microfoil, disks, 10mm, thinness 0.25μm, specific density 112.6μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, tube, 1000mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 200mm, diameter 2mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Titanium, microfoil, disks, 10mm, thinness 1.0μm, specific density 429μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, rod, 100mm, diameter 5mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 2mm, as drawn, 99.99+%
Titanium, tube, 200mm, outside diameter 6.35mm, inside diameter 5.53mm, wall thickness 0.41mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.1mm, inside diameter 5.1mm, wall thickness 0.5mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 3.0mm, annealed, 99.6+%
Titanium, microfoil, disks, 25mm, thinness 0.25μm, specific density 112.6μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, tube, 100mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Titanium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.99+%
Titanium, wire reel, 200m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.5mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.25mm, as drawn, 99.99+%
Titanium, tube, 500mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%