Merck
  • A whole-cell bioreporter approach for the genotoxicity assessment of bioavailability of toxic compounds in contaminated soil in China.
A whole-cell bioreporter approach for the genotoxicity assessment of bioavailability of toxic compounds in contaminated soil in China.
Environmental pollution (Barking, Essex : 1987) (2014-09-23)
Yizhi Song, Bo Jiang, Sicong Tian, Hui Tang, Zengjun Liu, Chuan Li, Jianli Jia, Wei E Huang, Xu Zhang, Guanghe Li
ABSTRACT
A whole-cell bacterial bioreporter Acinetobacter baylyi strain ADP1_recA_lux that responds to genotoxins was employed to directly assess the adverse effects of the bioavailable fraction of mitomycin C (MMC), benzo[a]pyrene (BaP), chromium (VI) and lead (II) in amended soils and soil samples from two fragile areas in China without soil pre-treatment. The amended soils containing pollutants with the concentrations as low as 0.4 mg/kg MMC, 0.5 mg/kg BaP, 520 mg/kg Cr (VI) and 2072 mg/kg Pb (II) were found to be toxic. Soil particle-associated pollutants accounted for 86%, 100%, 29%, and 92% of the genotoxicity in the MMC, BaP, Cr (VI), and Pb (II) amended soil, respectively. The soils from contaminated sites were also valid to be genotoxic. The results suggest both free and soil particle-associated pollutants are bioavailable to soil organisms and a solid-phase contact bioreporter assay to soil contamination could provide a rapid screening tool for environmental risk assessment.
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Chromium, microfoil, disks, 25mm, thinness 0.5μm, specific density 355μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, rod, 25mm, diameter 5.0mm, 99.7+%
Chromium, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, rod, 100mm, diameter 5.0mm, 99.7+%
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.03mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Chromium, rod, 25mm, diameter 2.0mm, 99.7+%
Chromium, microfoil, disks, 10mm, thinness 0.05μm, specific density 37.27μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, rod, 50mm, diameter 2.0mm, 99.7+%
Chromium, rod, 10mm, diameter 2.0mm, 99.7+%
Chromium, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Chromium, microfoil, disks, 10mm, thinness 0.025μm, specific density 18.21μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, microfoil, disks, 10mm, thinness 0.25μm, specific density 178.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.99+%
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Chromium, microfoil, disks, 25mm, thinness 0.25μm, specific density 178.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.04mm, permanent polyester support, 99.99+%
Chromium, microfoil, disks, 25mm, thinness 1.0μm, specific density 719μg/cm2, permanent mylar 3.5μm support, 99.99+%