Merck
  • Status of phthalate esters contamination in agricultural soils across China and associated health risks.
Status of phthalate esters contamination in agricultural soils across China and associated health risks.
Environmental pollution (Barking, Essex : 1987) (2014-09-10)
Lili Niu, Yang Xu, Chao Xu, Lingxiang Yun, Weiping Liu
ABSTRACT
The extensive utilization of phthalate-containing products has lead to ubiquitous contamination of phthalate esters (PAEs) in various matrices. However, comprehensive knowledge of their pollution in Chinese farmland and associated risks is still limited. In this study, 15 PAEs were determined in soils from agricultural fields throughout the Mainland China. The concentrations of Σ15PAEs were in the range of 75.0-6369 μg kg(-1). Three provinces (i.e., Fujian, Guangdong and Xinjiang, China) showed the highest loadings of PAEs. Bis(2-Ethylhexyl) phthalate (DEHP) was found as the most abundant component and contributed 71.5% to the ∑15PAEs. The major source of PAEs in arable soils was associated with the application of agricultural plastic films, followed by the activities for soil fertility. Furthermore, the non-cancer and carcinogenic risks of target PAEs were estimated. The hazard indexes (HIs) of PAEs in all samples were below 1 and the carcinogenic risk levels were all within 10(-4). Results from this study will provide valuable information for Chinese agricultural soil management and risk avoidance.
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Supelco
Phthalic acid, analytical standard
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Sigma-Aldrich
Benzyl benzoate, meets USP testing specifications
Sigma-Aldrich
Benzyl benzoate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Benzyl benzoate, natural, ≥99%, FG
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Benzyl benzoate, tested according to Ph. Eur.
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
USP
Benzyl benzoate, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzyl benzoate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Supelco
Benzyl benzoate, analytical standard
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl benzoate, ≥99%, FCC, FG