Merck
  • Clinical efficacy of mitoxantrone and Ara-C with or without etoposide salvage chemotherapy in adult patients with relapsed or refractory acute lymphoblastic leukemia: retrospective multicenter study of the Korean Adult ALL Working Party.
Clinical efficacy of mitoxantrone and Ara-C with or without etoposide salvage chemotherapy in adult patients with relapsed or refractory acute lymphoblastic leukemia: retrospective multicenter study of the Korean Adult ALL Working Party.
Acta haematologica (2014-08-30)
Jae-Sook Ahn, Deok-Hwan Yang, Sung-Hoon Jung, Je-Jung Lee, Inho Kim, Seonyang Park, Joo Seop Chung, Ho-Jin Shin, Dae-Young Kim, Kyoo-Hyung Lee, Joon Ho Moon, Sang Kyun Sohn, Ik-Chan Song, Deog-Yeon Jo, Young Don Joo
ABSTRACT
Mitoxantrone is a conventional agent for relapsed or refractory acute lymphoblastic leukemia (ALL). However, an effective combination with other drugs and a feasible dosage has not been identified. A retrospective study of 46 patients with relapsed or refractory ALL was conducted to determine the efficacy of mitoxantrone and Ara-C treatment with (MEC) and or without etoposide (MC). Twenty-seven and 19 patients received MC and MEC chemotherapy, respectively. Twenty-two (48%) patients showed overall response [complete response (CR), 33%; CR with incomplete platelet recovery (CRp), 15%], and 10 of 22 responders received allogeneic stem cell transplantation (SCT). Median overall survival (OS) was 6.2 months (95% confidence interval, 3.41-9.0). Thirteen (48%) patients in the MC group and 9 (47%) in the MEC group achieved CR/CRp (p = 0.96). Treatment-related mortalities in the MC and MEC groups were 3 (11%) and 4 (21%), respectively (p = 0.36). However, the MEC group frequently presented with grade 3 or higher bacteremia/candidemia (p = 0.013). No difference in OS was observed between the two groups (p = 0.769). In conclusion, salvage therapy consisting of mitoxantrone and Ara-C without etoposide appeared to be an effective bridge therapy to allogeneic SCT for patients with refractory or relapsed ALL.
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitoxantrone dihydrochloride, ≥97% (HPLC)
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Mitoxantrone hydrochloride, European Pharmacopoeia (EP) Reference Standard