Merck
  • EGF regulates claudin-2 and -4 expression through Src and STAT3 in MDCK cells.
EGF regulates claudin-2 and -4 expression through Src and STAT3 in MDCK cells.
Journal of cellular physiology (2014-06-10)
Vicky García-Hernández, Catalina Flores-Maldonado, Ruth Rincon-Heredia, Odette Verdejo-Torres, José Bonilla-Delgado, Ivan Meneses-Morales, Patricio Gariglio, Rubén G Contreras
ABSTRACT
Epidermal Growth Factor (EGF) is a key regulator of epithelial paracellular permeability, a property that depends on tight junctions (TJ) and can be evaluated through the measurement of the transepithelial electrical resistance (TER). EGF increases the TER of MDCK monolayers by inducing ERK1/2-dependent downregulation of claudin-2 (CLDN-2) and upregulation of claudin-4 (CLDN-4). Because either increments or decrements in TER often involve Src activation and epithelial cell differentiation occasionally depends on STAT3, here we investigated whether EGF might control CLDN-2 downregulation and CLDN-4 upregulation through those proteins. We found that EGF induces Src activation necessary for the reduction of CLDN-2 at the TJ, the degradation of this CLDN, the reduction of the cellular levels of its mRNA and the resulting increase of TER. EGF-induced changes on CLDN-2 protein and mRNA also depend on STAT3 activity. This growth factor increases the levels of STAT3 phosphorylated at Y705 in the nucleus, a process that depends on Src activation. Interestingly, Src and STAT3 activation do not exclusively mediate the EGF-induced downregulation of CLDN-2, but they are also implicated in the EGF-induced CLDN-4 transcription, translation, and exocytic fusion into TJ. Our results indicate that EGF controls the levels of CLDN-2 and -4 proteins and mRNAs through Src and STAT3 activity.
Sigma-Aldrich
Gö 6983, A potent, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase C (PKC) that inhibits several PKC isozymes.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)