Merck
  • Neural cell adhesion molecule expression, neuroendocrine differentiation and prognosis in lung carcinoma.
Neural cell adhesion molecule expression, neuroendocrine differentiation and prognosis in lung carcinoma.
European journal of cancer (Oxford, England : 1990) (1991-01-01)
R E Kibbelaar, K E Moolenaar, R J Michalides, P C Van Bodegom, R G Vanderschueren, S S Wagenaar, K P Dingemans, D Bitter-Suermann, O Dalesio, N Van Zandwijk
ABSTRACT
We investigated the expression of the neural cell adhesion molecule (NCAM) in a series of surgically resected lung carcinomas of various histological subtypes by means of a panel of monoclonal antibodies recognising different N-CAM epitopes. In a subgroup of 56 tumours, the results of immunostaining with MAb 123C3--the antibody studied most extensively in our material--were compared to the ultrastructure, and in 231 radically resected non-small cell carcinomas, with histological tumour type and with clinical follow-up data. N-CAM expression was not limited to neuroendocrine tumours, as assessed ultrastructurally. Non-small cell lung carcinomas positive for MAb 123C3 showed post-operative overall and disease-free survival times significantly shorter than 123C3-negative non-small cell carcinomas.
Sigma-Aldrich
CD56 (123C3.D5) Mouse Monoclonal Antibody, clone 123C3.D5, unconjugated, Cell Marque
Sigma-Aldrich
CD56 (MRQ-42) Rabbit Monoclonal Antibody