Merck
  • Enhanced expression of programmed death-1 (PD-1)/PD-L1 in salivary glands of patients with Sjögren's syndrome.
Enhanced expression of programmed death-1 (PD-1)/PD-L1 in salivary glands of patients with Sjögren's syndrome.
The Journal of rheumatology (2005-11-03)
Masaya Kobayashi, Seiji Kawano, Saori Hatachi, Chiyo Kurimoto, Taku Okazaki, Yoshiko Iwai, Tasuku Honjo, Yoshimasa Tanaka, Nagahiro Minato, Takahide Komori, Sakan Maeda, Shunichi Kumagai
ABSTRACT
Programmed death-1 (PD-1) mediates a negative signal and introduces tolerance for lymphocytes. Dysfunction of the PD-1 pathway is thought to result in autoimmune diseases such as rheumatoid arthritis (RA). To investigate the role of the PD-1/PD-L system in the pathology of Sjögren's syndrome (SS), we examined the expression of PD-1 and its ligand PD-L1 in salivary lymphocytes and salivary glands from patients with SS. Flow cytometry analysis was used to determine expression of PD-1 in SS salivary lymphocytes. Intracellular staining of interleukin 10 (IL-10) was performed after stimulation with PMA and ionomycin. Indirect immunohistochemistry was used to investigate the expression of PD-1 and PD-L1. The mean fluorescence intensity of PD-1 expression in SS salivary lymphocytes was significantly higher than that from healthy controls and patients with RA or systemic lupus erythematosus. PD-1-positive SS salivary lymphocytes expressed IL-10 intracellularly upon PMA/ionomycin stimulation. Immunohistochemical analysis showed that PD-1 was expressed on infiltrating lymphocytes in salivary gland from 52% of SS patients, and PD-L1 was expressed on ductal and acinar epithelial cells from 68% of SS patients. In vitro analysis using HSG cells revealed that PD-L1 was induced by interferon-gamma but not by tumor necrosis factor-alpha and IL-1beta. PD-1 is expressed on T lymphocytes and PD-L1 on epithelial cells from inflamed salivary glands of patients with SS, which suggests that dysfunction of the PD-1/PD-L1 pathway may be related to tolerance for lymphocytes, which causes SS.
Sigma-Aldrich
PD-1 (NAT105) Mouse Monoclonal Antibody