Merck
Estrogen-mediated regulation of mitochondrial gene expression.
Molecular endocrinology (Baltimore, Md.) (2014-11-07)
Maria I G Lopez Sanchez, Anne-Marie J Shearwood, Tiongsun Chia, Stefan M K Davies, Oliver Rackham, Aleksandra Filipovska
ABSTRACT
Estrogens, in particular 17β-estradiol, are well-known regulators of essential cellular functions; however, discrepancies remain over the mechanisms by which they act on mitochondria. Here we propose a novel mechanism for the direct regulation of mitochondrial gene expression by estrogen under metabolic stress. We show that in serum-depleted medium, estrogen stimulates a rapid relocation of estrogen receptor-α to mitochondria, in which it elicits a cellular response, resulting in an increase in mitochondrial RNA abundance. Mitochondrial RNA levels are regulated through the association of estrogen receptor-α with 17β-hydroxysteroid dehydrogenase 10, a multifunctional protein involved in steroid metabolism that is also a core subunit of the mitochondrial ribonuclease P complex responsible for the cleavage of mitochondrial polycistronic transcripts. Processing of mitochondrial transcripts affects mitochondrial gene expression by controlling the levels of mature RNAs available for translation. This work provides the first mechanism linking RNA processing and estrogen activation in mitochondrial gene expression and underscores the coordinated response between the nucleus and mitochondria in response to stress.
Sigma-Aldrich
Anti-ELAC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HSD17B10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PTCD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution