Merck
  • Trafficking of TRPP2 by PACS proteins represents a novel mechanism of ion channel regulation.
Trafficking of TRPP2 by PACS proteins represents a novel mechanism of ion channel regulation.
The EMBO journal (2005-02-05)
Michael Köttgen, Thomas Benzing, Thomas Simmen, Robert Tauber, Björn Buchholz, Sylvain Feliciangeli, Tobias B Huber, Bernhard Schermer, Albrecht Kramer-Zucker, Katja Höpker, Katia Carmine Simmen, Christoph Carl Tschucke, Richard Sandford, Emily Kim, Gary Thomas, Gerd Walz
ABSTRACT
The trafficking of ion channels to the plasma membrane is tightly controlled to ensure the proper regulation of intracellular ion homeostasis and signal transduction. Mutations of polycystin-2, a member of the TRP family of cation channels, cause autosomal dominant polycystic kidney disease, a disorder characterized by renal cysts and progressive renal failure. Polycystin-2 functions as a calcium-permeable nonselective cation channel; however, it is disputed whether polycystin-2 resides and acts at the plasma membrane or endoplasmic reticulum (ER). We show that the subcellular localization and function of polycystin-2 are directed by phosphofurin acidic cluster sorting protein (PACS)-1 and PACS-2, two adaptor proteins that recognize an acidic cluster in the carboxy-terminal domain of polycystin-2. Binding to these adaptor proteins is regulated by the phosphorylation of polycystin-2 by the protein kinase casein kinase 2, required for the routing of polycystin-2 between ER, Golgi and plasma membrane compartments. Our paradigm that polycystin-2 is sorted to and active at both ER and plasma membrane reconciles the previously incongruent views of its localization and function. Furthermore, PACS proteins may represent a novel molecular mechanism for ion channel trafficking, directing acidic cluster-containing ion channels to distinct subcellular compartments.