Merck
  • Dielectrophoretic technique for measurement of chemical and biological interactions.
Dielectrophoretic technique for measurement of chemical and biological interactions.
Analytical chemistry (2009-08-12)
Sang Hyun Baek, Woo-Jin Chang, Ju-Yeoul Baek, Dae Sung Yoon, Rashid Bashir, Sang Woo Lee
ABSTRACT
We present a novel dielectrophoretic technique that can be used to characterize molecular interactions inside a microfluidic device. Our approach allows functionalized beads which are initially at rest on a functionalized surface to be pulled away from the surface by the dielectrophoretic force acting on the beads. As a result, the interaction between the molecules on the surface and the beads can be quantitatively examined. We report detailed experimental results and validate the results with a model to show that the technique can be used to measure forces of interaction between molecules under various experimental conditions.
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)