Merck
  • Effective purification of single-walled carbon nanotubes with reversible noncovalent functionalization.
Effective purification of single-walled carbon nanotubes with reversible noncovalent functionalization.
Langmuir : the ACS journal of surfaces and colloids (2010-01-06)
Fushen Lu, Xin Wang, Mohammed J Meziani, Li Cao, Leilei Tian, Matthew A Bloodgood, Jackie Robinson, Ya-Ping Sun
ABSTRACT
An effective purification method for single-walled carbon nanotubes (SWNTs) based on a combination of oxidative acid treatment and reversible noncovalent functionalization with 1-pyreneacetic acid is reported. The functionalization was selective toward the nanotubes, allowing a nearly complete removal of residual metal catalysts and carbonaceous impurities. The resulting highly pure SWNTs remained solvent-dispersible, a valuable feature to potential applications that require solvent-based processing. The functionalization agent could be recovered quantitatively and reused. Effects of the purification process on the composition and properties of the nanotube sample were evaluated.
Sigma-Aldrich
1-Pyreneacetic acid, 97%