Merck
  • Pharmacological relevance of endoxifen in a laboratory simulation of breast cancer in postmenopausal patients.
Pharmacological relevance of endoxifen in a laboratory simulation of breast cancer in postmenopausal patients.
Journal of the National Cancer Institute (2014-09-27)
Philipp Y Maximov, Russell E McDaniel, Daphne J Fernandes, Puspanjali Bhatta, Valeriy R Korostyshevskiy, Ramona F Curpan, V Craig Jordan
ABSTRACT
Tamoxifen is metabolically activated via a CYP2D6 enzyme system to the more potent hydroxylated derivatives 4-hydroxytamoxifen and endoxifen. This study addresses the pharmacological importance of endoxifen by simulating clinical scenarios in vitro. Clinical levels of tamoxifen metabolites in postmenopausal breast cancer patients previously genotyped for CYP2D6 were used in vitro along with clinical estrogen levels (estrone and estradiol) in postmenopausal patients determined in previous studies. The biological effects on cell growth were evaluated in a panel of estrogen receptor-positive breast cancer cell lines via cell proliferation assays and real-time polymerase chain reaction (PCR). Data were analyzed with one- and two-way analysis of variance and Student's t test. All statistical tests were two-sided. Postmenopausal levels of estrogen-induced proliferation of all test breast cancer cell lines (mean fold induction ± SD vs vehicle control: MCF-7 = 11 ± 1.74, P < .001; T47D = 7.52 ± 0.72, P < .001; BT474 = 1.75 ± 0.23, P < .001; ZR-75-1 = 5.5 ± 1.95, P = .001. Tamoxifen and primary metabolites completely inhibited cell growth regardless of the CYP2D6 genotype in all cell lines (mean fold induction ± SD vs vehicle control: MCF-7 = 1.57 ± 0.38, P = .54; T47D = 1.17 ± 0.23, P = .79; BT474 = 0.96 ± 0.2, P = .98; ZR-75-1 = 0.86 ± 0.67, P = .99). Interestingly, tamoxifen and its primary metabolites were not able to fully inhibit the estrogen-stimulated expression of estrogen-responsive genes in MCF-7 cells (P < .05 for all genes), but the addition of endoxifen was able to produce additional antiestrogenic effect on these genes. The results indicate that tamoxifen and other metabolites, excluding endoxifen, completely inhibit estrogen-stimulated growth in all cell lines, but additional antiestrogenic action from endoxifen is necessary for complete blockade of estrogen-stimulated genes. Endoxifen is of supportive importance for the therapeutic effect of tamoxifen in a postmenopausal setting.
Product Description
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Supelco
Estrone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Estrone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Tamoxifen citrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Tamoxifen, analytical standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®