Merck
  • Artificial receptors for the extraction of nucleoside metabolite 7-methylguanosine from aqueous media made by molecular imprinting.
Artificial receptors for the extraction of nucleoside metabolite 7-methylguanosine from aqueous media made by molecular imprinting.
Journal of chromatography. A (2014-09-15)
Aleksandra Krstulja, Coralie De Schutter, Patrick Favetta, Panagiotis Manesiotis, Luigi A Agrofoglio
ABSTRACT
A series of imprinted polymers targeting nucleoside metabolites, prepared using a template analogue approach, are presented. These were prepared following selection of the optimum functional monomer by solution association studies using (1)H NMR titrations whereby methacrylic acid was shown to be the strongest receptor with and affinity constant of 621±51Lmol(-1)vs. 110±16Lmol(-1) for acrylamide. The best performing polymers were prepared using methanol as porogenic co-solvent and although average binding site affinities were marginally reduced, 2.3×10(4)Lmol(-1)vs. 2.7×10(4)Lmol(-1) measured for a polymer prepared in acetonitrile, these polymers contained the highest number of binding sites, 5.27μmolg(-1)vs. 1.64μmolg(-1), while they also exhibited enhanced selectivity for methylated guanosine derivatives. When applied as sorbents in the extraction of nucleoside derivative cancer biomarkers from synthetic urine samples, significant sample clean-up and recoveries of up to 90% for 7-methylguanosine were achieved.
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Water, for TOC analysis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Sodium acrylate, 97%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Water, suitable for ion chromatography
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C