Merck
  • Enzymatic synthesis of multi-component copolymers and their structural characterization.
Enzymatic synthesis of multi-component copolymers and their structural characterization.
Molecular diversity (2004-04-08)
Rajesh Kumar, Rahul Tyagi, Virinder S Parmar, Lynne A Samuelson, Jayant Kumar, Arthur C Watterson
ABSTRACT
The use of enzymes in synthetic applications has increased dramatically in the recent years and the field of polymer science is part of this trend. Synthesis of a variety of polymers using lipase catalyzed (Candida antarctica) polymerization reactions has led to a variety of new materials with interesting properties in our laboratories. This paper describes the synthesis of multi-component polyesters and mixed polymers having polyester and polyamide linkages under solvent-less conditions using Candida antarctica lipase B. The effect of a third component, i.e. a series of 1,omega-alkanediols (1,4-butanediol, 1,6-hexanediol, 1,8-octanediol, 1,10-decanediol, 1,12-dodecanediol, 1,14-tetradecanediol and 1,16-hexadecanediol) on the copolymerization reaction of dimethyl 5-hydroxyisophthalate with poly(ethylene glycol 600) has been studied and the mechanism for the incorporation of the third component is proposed. We have also studied the effect of different functional groups during terpolymerization reaction of dimethyl 5-hydroxyisophthalate with poly(ethylene glycol) by adding a third component having different functionalities (1,6-hexanediol, 1,6-hexanediamine or 1,6-hexanedithiol) and compared the effect of hydroxyl, amine and thiol groups on the polymerization reactions.
Sigma-Aldrich
Dimethyl 5-hydroxyisophthalate, 98%