Merck
General synthesis of thiophene and selenophene-based heteroacenes.
Organic letters (2005-11-05)
Toshihiro Okamoto, Kenichi Kudoh, Atsushi Wakamiya, Shigehiro Yamaguchi
ABSTRACT
[reaction: see text] A new intramolecular triple cyclization of bis(o-haloaryl)diacetylenes, via dilithiation followed by reaction with chalcogen elements, produces pi-conjugated compounds containing heterole-1,2-dichalcogenin-heterole fused tricyclic skeletons. The subsequent dechalcogenation with copper metal affords a series of thiophene- and selenophene-based heteroacenes.
Sigma-Aldrich
Selenophene, 97%