Merck
  • One-step synthesis of heterocyclic privileged medicinal scaffolds by a multicomponent reaction of malononitrile with aldehydes and thiols.
One-step synthesis of heterocyclic privileged medicinal scaffolds by a multicomponent reaction of malononitrile with aldehydes and thiols.
The Journal of organic chemistry (2007-04-06)
Nikolai M Evdokimov, Artem S Kireev, Andrey A Yakovenko, Mikhail Yu Antipin, Igor V Magedov, Alexander Kornienko
ABSTRACT
Heterocyclic privileged medicinal scaffolds involving pyridine, 1,4-dihydropyridine, chromeno[2,3-b]pyridine, and dihydro-1,4-dithiepine frameworks are prepared via a single-step multicomponent reaction of structurally diverse aldehydes with various thiols and malononitrile. Mechanistic studies of the synthetic pathway leading to pyridines reveal that 1,4-dihydropyridines undergo oxidation by the intermediate Knoevenagel adducts rather than by air oxygen. The use of o,o'-disubstituted aromatic aldehydes leads to the corresponding 1,4-dihydropyridines, whereas salicylic aldehydes result in chromeno[2,3-b]pyridines. Reactions of ethanedithiol as a thiol component produce dimeric pyridines with sterically unencumbered aldehydes, while o,o'-disubstituted aromatic aldehydes give dihydro-1,4-dithiepines. Thus, depending on the aldehyde and thiol types, diverse libraries of medicinally relevant compounds can be prepared by a simple one-step process involving no chromatography.
Sigma-Aldrich
Malononitrile, Arxada quality, ≥99.0% (calculated, GC, KF)
Sigma-Aldrich
Malononitrile, ≥99%