Merck
  • Contrasting patterns of allochthony among three major groups of crustacean zooplankton in boreal and temperate lakes.
Contrasting patterns of allochthony among three major groups of crustacean zooplankton in boreal and temperate lakes.
Ecology (2014-08-29)
Martin Berggren, Susan E Ziegler, Nicolas F St-Gelais, Beatrix E Beisner, Paul A Del Giorgio
ABSTRACT
The importance of terrestrial-derived organic matter for lake zooplankton communities remains debated, partly because little is known about the basic pathways by which allochthonous carbon is transferred to zooplankton, and whether these vary among the major taxonomic and functional groups. We quantified allochthony of three zooplankton groups (Cladocera, Calanoida, and Cyclopoida) across 18 lakes in Quebec, spanning broad gradients of dissolved organic matter (DOM) and lake trophy, using a multi-isotope (delta2H + delta13C), multi-source (terrestrial, phytoplanktonic, benthic) approach. All three zooplankton groups had significant levels of allochthony, but differed greatly in their respective patterns across lakes. Allochthony in Calanoida and Cyclopoida was linked to detrital food chains based on particulate organic matter (POM) and on DOM, respectively, whereas in Cladocera it appeared related to both pathways; not surprisingly this latter group had the highest mean allochthony (0.31; compared to 0.18 in Cyclopoida and 0.16 in Calanoida). This study highlights the complexity of the pathways of delivery and transfer of terrestrial organic matter in freshwaters, and underscores the role that microbial food webs play in this transfer.
Product Description
Supelco
Water, for TOC analysis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Deuterium, 99.8 atom % D