Merck
  • Role of FDG-PET in the implementation of involved-node radiation therapy for Hodgkin lymphoma patients.
Role of FDG-PET in the implementation of involved-node radiation therapy for Hodgkin lymphoma patients.
International journal of radiation oncology, biology, physics (2014-07-19)
Théodore Girinsky, Anne Aupérin, Vincent Ribrag, Manel Elleuch, Christophe Fermé, Guillaume Bonniaud, Claude Ruelle, Jean-Louis Alberini, Aljosa Celebic, Véronique Edeline
ABSTRACT
This study examines the role of (18)F-labeled fluorodeoxyglucose positron emission tomography (FDG-PET) in the implementation of involved-node radiation therapy (INRT) in patients treated for clinical stages (CS) I/II supradiaphragmatic Hodgkin lymphoma (HL). Patients with untreated CS I/II HL enrolled in the randomized EORTC/LYSA/FIL Intergroup H10 trial and participating in a real-time prospective quality assurance program were prospectively included in this study. Data were electronically obtained from 18 French cancer centers. All patients underwent APET-computed tomography (PET-CT) and a post-chemotherapy planning CT scanning. The pre-chemotherapy gross tumor volume (GTV) and the postchemotherapy clinical target volume (CTV) were first delineated on CT only by the radiation oncologist. The planning PET was then co-registered, and the delineated volumes were jointly analyzed by the radiation oncologist and the nuclear medicine physician. Lymph nodes undetected on CT but FDG-avid were recorded, and the previously determined GTV and CTV were modified according to FDG-PET results. From March 2007 to February 2010, 135 patients were included in the study. PET-CT identified at least 1 additional FDG-avid lymph node in 95 of 135 patients (70.4%; 95% confidence interval [CI]: 61.9%-77.9%) and 1 additional lymph node area in 55 of 135 patients (40.7%; 95% CI: 32.4%-49.5%). The mean increases in the GTV and CTV were 8.8% and 7.1%, respectively. The systematic addition of PET to CT led to a CTV increase in 60% of the patients. Pre-chemotherapy FDG-PET leads to significantly better INRT delineation without necessarily increasing radiation volumes.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
2-Fluoro-2-deoxy-D-glucose, glycosylation inhibitor, glucose analog
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Bleomycin sulfate, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard