Merck
  • Respiratory symptoms and lung function abnormalities related to work at a flavouring manufacturing facility.
Respiratory symptoms and lung function abnormalities related to work at a flavouring manufacturing facility.
Occupational and environmental medicine (2014-06-04)
Kristin J Cummings, Randy J Boylstein, Marcia L Stanton, Chris A Piacitelli, Nicole T Edwards, Ryan F LeBouf, Kathleen Kreiss
ABSTRACT
To better understand respiratory symptoms and lung function in flavouring manufacturing workers. We offered a questionnaire and lung function testing to the current workforce of a flavouring manufacturing facility that had transitioned away from diacetyl and towards substitutes in recent years. We examined symptoms, spirometric parameters and diffusing capacity measurements by exposure variables, including facility tenure and time spent daily in production areas. We used linear and logistic regression to develop final models adjusted for age and smoking status. A total of 367 (93%) current workers participated. Shortness of breath was twice as common in those with tenure ≥ 7 years (OR 2.0, 95% CI 1.1 to 3.6). Other chest symptoms were associated with time spent daily in production. Participants who spent ≥ 1 h daily in production areas had twice the odds of any spirometric abnormality (OR 2.3; 95% CI 1.1 to 5.3) and three times the odds of low diffusing capacity (OR 2.8; 95% CI 0.9 to 9.4) than other participants. Mean spirometric parameters were significantly lower in those with tenure ≥ 7 years and those who spent ≥ 1 h daily in production. Mean diffusing capacity parameters were significantly lower in those with tenure ≥ 7 years. Differences in symptoms and lung function could not be explained by age, smoking status or employment at another flavouring plant. Symptoms and lung function findings were consistent with undiagnosed or subclinical obliterative bronchiolitis and associated with workplace exposures. Further efforts to lower exposures to flavouring chemicals, including diacetyl substitutes, are warranted.
Sigma-Aldrich
2,3-Butanedione, 97%
Supelco
2,3-Butanedione, analytical standard