- Identification of erythroferrone as an erythroid regulator of iron metabolism.
Identification of erythroferrone as an erythroid regulator of iron metabolism.
Nature genetics (2014-06-02)
Léon Kautz, Grace Jung, Erika V Valore, Stefano Rivella, Elizabeta Nemeth, Tomas Ganz
PMID24880340
ABSTRACT
Recovery from blood loss requires a greatly enhanced supply of iron to support expanded erythropoiesis. After hemorrhage, suppression of the iron-regulatory hormone hepcidin allows increased iron absorption and mobilization from stores. We identified a new hormone, erythroferrone (ERFE), that mediates hepcidin suppression during stress erythropoiesis. ERFE is produced by erythroblasts in response to erythropoietin. ERFE-deficient mice fail to suppress hepcidin rapidly after hemorrhage and exhibit a delay in recovery from blood loss. ERFE expression is greatly increased in Hbb(th3/+) mice with thalassemia intermedia, where it contributes to the suppression of hepcidin and the systemic iron overload characteristic of this disease.
Sigma-Aldrich
Erythropoietin human, EPO, recombinant, expressed in HEK 293 cells, suitable for cell culture