Merck
  • Relative vitamin A values of 9-cis- and 13-cis-β-carotene do not differ when fed at physiological levels during vitamin A depletion in Mongolian gerbils (Meriones unguiculatus).
Relative vitamin A values of 9-cis- and 13-cis-β-carotene do not differ when fed at physiological levels during vitamin A depletion in Mongolian gerbils (Meriones unguiculatus).
The British journal of nutrition (2014-04-09)
Kara A Bresnahan, Christopher R Davis, Sherry A Tanumihardjo
ABSTRACT
Provitamin A biofortification of staple crops may decrease the prevalence of vitamin A (VA) deficiency if widely adopted in target countries. To assess the impact of processing methods on the VA value of plant foods, the unique bioefficacies of cis-βC isomers (formed during cooking) compared with all-trans (at) β-carotene (βC) must be determined. The bioefficacies of 9-cis (9c)- and 13-cis (13c)-βC isomers were compared with those of the at-βC isomer and VA positive (VA+) and negative (VA - ) controls in VA-depleted Mongolian gerbils (Meriones unguiculatus) in two experimental studies (study 1, n 56; study 2, n 57). A 3- or 4-week depletion period was followed by a 3- or 4-week treatment period in which the groups received oral doses of the 9c-, 13c- or at-βC isomers in cottonseed oil (study 1, 15 nmol/d; study 2, 30 nmol/d). In study 1, the βC isomers did not maintain baseline liver VA stores in all groups (0.69 (SD 0.20) μmol/liver) except in the VA+group (0.56 (SD 0.10) μmol/liver) (P= 0.0026). The βC groups were similar to the VA+group, but the 9c- and 13c-βC groups did not differ from the VA - group (0.39 (SD 0.09) μmol/liver). In study 2, the βC isomers maintained baseline liver VA stores in all the βC groups (0.35 (SD 0.13) μmol/liver), and in the VA+group, the VA supplement (0.54 (SD 0.19) μmol/liver) exceeded the baseline VA status (0.38 (SD 0.15) μmol/liver) (P< 0.0001); however, the 9c-βC group did not differ from the VA - group (0.20 (SD 0.07) μmol/liver). In vivo isomerisation of βC was confirmed in both experimental studies. Lower VA bioconversion factor values were obtained for the cis-βC isomers in study 2 when compared with study 1, but higher values were obtained for the at-βC isomer. Dose and VA status clearly affect bioconversion factors. In conclusion, the cis-βC isomers yielded similar liver VA stores to the at-βC isomer in Mongolian gerbils, and liver VA stores of the 9c- and 13c-βC groups did not differ when the doses were provided at physiological levels over time in two studies.
Sigma-Aldrich
β-Carotene, synthetic, ≥93% (UV), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Pricing and availability is not currently available.
Sigma-Aldrich
β-Carotene, synthetic, ≥95% (HPLC), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Pricing and availability is not currently available.
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Pricing and availability is not currently available.
Supelco
β-Carotene, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Retinyl acetate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
Pricing and availability is not currently available.
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Pricing and availability is not currently available.
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Pricing and availability is not currently available.
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Retinyl acetate, United States Pharmacopeia (USP) Reference Standard, solution of retinyl acetate (vitamin A) in peanut oil
Pricing and availability is not currently available.