Merck
  • Protein arginine methyltransferase 6 enhances ligand-dependent and -independent activity of estrogen receptor α via distinct mechanisms.
Protein arginine methyltransferase 6 enhances ligand-dependent and -independent activity of estrogen receptor α via distinct mechanisms.
Biochimica et biophysica acta (2014-04-20)
Yang Sun, Hwa Hwa Chung, Amanda Rui En Woo, Valerie C-L Lin
ABSTRACT
Recent studies reported that protein arginine methyltransferase 6 (PRMT6) enhances estrogen-induced activity of estrogen receptor α (ERα) and dysfunction of PRMT6 is associated with overall better survival for ERα-positive breast cancer patients. However, it is unclear how PRMT6 promotes ERα activity. Here we report that PRMT6 specifically interacts with ERα at its ligand-binding domain. PRMT6 also methylates ERα both in vitro and in vivo. In addition to enhancing estrogen-induced ERα activity, PRMT6 over-expression up-regulates estrogen-independent activity of ERα and PRMT6 gene silencing in MCF7 cells inhibits ligand-independent ERα activation. More interestingly, the effect of PRMT6 on the ligand-independent ERα activity does not require its methyltransferase activity. Instead, PRMT6 competes with Hsp90 for ERα binding: PRMT6 and Hsp90 bindings to ERα are mutually exclusive and PRMT6 over-expression reduces ERα interaction with Hsp90. In conclusion, PRMT6 requires its methyltransferase activity to enhance ERα's ligand-induced activity, but its effect on ligand-independent activity is likely mediated through competing with Hsp90 for binding to the C-terminal domain of ERα. PRMT6-ERα interaction would prevent ERα-Hsp90 association. Since Hsp90 and associated chaperones serve to maintain ERα conformation for ligand-binding yet functionally inactive, inhibition of ERα-Hsp90 interaction would relieve ERα from the constraint of chaperone complex.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Arginine, European Pharmacopoeia (EP) Reference Standard