Merck
  • Exploitation of starch industry liquid by-product to produce bioactive peptides from rice hydrolyzed proteins.
Exploitation of starch industry liquid by-product to produce bioactive peptides from rice hydrolyzed proteins.
Food chemistry (2014-03-07)
Lucilla Dei Piu', Annalisa Tassoni, Diana Isabella Serrazanetti, Maura Ferri, Elena Babini, Davide Tagliazucchi, Andrea Gianotti
ABSTRACT
Small peptides show higher antioxidant capacity than native proteins and may be absorbed in the intestine without further digestion. In our study, a protein by-product from rice starch industry was hydrolyzed with commercial proteolytic enzymes (Alcalase, Neutrase, Flavourzyme) and microbial whole cells of Bacillus spp. and the released peptides were tested for antioxidant activity. Among enzymes, Alcalase was the most performing, while microbial proteolytic activity was less efficient. Conversely, the antioxidant activity was higher in the samples obtained by microbial hydrolysis and particularly with Bacillus pumilus AG1. The sequences of low molecular weight antioxidant peptides were determined and analyzed for aminoacidic composition. The results obtained so far suggest that the hydrolytic treatment of this industrial by-product, with selected enzymes and microbial systems, can allow its exploitation for the production of functional additives and supplements rich in antioxidant peptides, to be used in new food formulas for human consumption.
Sigma-Aldrich
Starch from wheat, Unmodified
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch from potato, Powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch from potato, Soluble
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch from rice
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch from corn
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch from corn, practical grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Sigma-Aldrich
Starch from corn, Unmodified waxy corn starch of essentially pure amylopectin; contains only trace amounts of amylose.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
€104.00