Merck
  • Synthesis and characterization of new derivatives of alginic acid and evaluation of their iron(III)-crosslinked beads as potential controlled release matrices.
Synthesis and characterization of new derivatives of alginic acid and evaluation of their iron(III)-crosslinked beads as potential controlled release matrices.
Pharmaceutical development and technology (2013-09-17)
Samer R Abulateefeh, Mohammad A Khanfar, Ramia Z Al Bakain, Mutasem O Taha
ABSTRACT
The excellent gelling and safety profiles of alginic acid combined, however, with drawbacks of its ionotropically crosslinked beads (i.e. their quick release of loaded drugs) prompted us to chemically modify alginic acid. Alginic acid was chemically conjugated with four amines of varying hydrophilic-hydrophobic properties (i.e. tris(hydroxymethyl)methyl-, allyl-, benzyl- or pentyl-amines) in an attempt to enhance the drug release profiles from respective metal crosslinked beads. Chemical conjugation procedures were performed using dicyclohexylcarbodiimide as a coupling agent and the resulting new derivatives were characterized using proton nuclear magnetic resonance ((1)H NMR), infrared (IR) spectroscopy and differential scanning calorimetry (DSC). These modified polymers were used to prepare iron (III)-crosslinked beads loaded with folic acid as model drug, which were tested in vitro to assess their folic acid release profiles. Interestingly, the resulting beads accessed enteric release kinetics, with tris(hydroxymethyl)methyl-amide alginic conjugate producing most pronounced enteric profile. The results suggest the possibility of achieving controlled drug release from alginate-based beads via facile chemical modification of alginic acid.
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Alginic acid sodium salt, powder
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1m, diameter 0.5mm, as drawn, 99.998%
Iron, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, rod, 500mm, diameter 12.7mm, as drawn, 99.95%
Iron, foil, 50x50mm, thickness 2mm, as rolled, 99.99+%
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, rod, 200mm, diameter 25mm, as drawn, 98+%
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Iron, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 300x300mm, thickness 2.0mm, as rolled, armco« soft ingot 99.8%
Iron, wire reel, 1m, diameter 0.025mm, as drawn, 99.99+%
Iron, foil, 300x300mm, thickness 0.15mm, hard, 99.5%
Iron, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Iron, wire reel, 1000m, diameter 0.125mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.005mm, 99.85%
Iron, tube, 500mm, outside diameter 3.18mm, inside diameter 1.96mm, wall thickness 0.61mm, as drawn, 99.5%
Iron, tube, 200mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.5%