Merck
  • Suitability of several carbon sorbents for the fractionation of various sub-groups of toxic polychlorinated biphenyls, polychlorinated dibenzo-p-dioxins and polychlorinated dibenzofurans.
Suitability of several carbon sorbents for the fractionation of various sub-groups of toxic polychlorinated biphenyls, polychlorinated dibenzo-p-dioxins and polychlorinated dibenzofurans.
Journal of chromatography. A (2001-06-14)
M Concejero, L Ramos, B Jiménez, B Gómara, E Abad, J Rivera, M J González
ABSTRACT
Feasibility of several sorbents, Amoco PX-21, Carbosphere, Carbopack B and C and 2-(1-pyrenyl) ethyldimethylsilylated silica gel (PYE), for the fractionation of polychlorinated biphenyls, polychlorinated dibenzo-p-dioxins and furans was investigated. Selection was based on their suitability for quantitative isolation of the target compounds with special attention for the most toxic mono- and non-ortho-CBs to prevent coelution with other congeners during the final determination by gas chromatography. Cost-effectiveness in terms of solvent and time consumption as well as feasibility for routine analysis and automation were considered additional merits of the methods compared. Final evaluation of the procedures providing the best results was done by comparison of the results obtained from the analysis of real-life samples. The results showed that, among the sorbents tested, Carbopack B and PYE were the most suitable for routine analysis. In particular, these sorbents allowed a more reliable determination of the toxic congeners and, consequently, of the toxic equivalents of 2,3,7,8-tetrachlorodibenzo-p-dioxin content in environmental samples.
Product Description
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Supelco
Carbon Adsorbent Sampler Kit, matrix CMS/SGPC Kit I
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea