Merck
  • Experimental uses of flunixin meglumine and phenylbutazone in food-producing animals.
Experimental uses of flunixin meglumine and phenylbutazone in food-producing animals.
Journal of the American Veterinary Medical Association (1989-01-01)
M Kopcha, A S Ahl
ABSTRACT
Presently, in the United States, there are no nonsteroidal anti-inflammatory drugs, except aspirin, that are approved for use in animals intended for food production. Use of phenylbutazone, flunixin meglumine, and dipyrone for treatment of food animals may be considered in special circumstances. Such use requires strict adherence to FDA guidelines for extra-label use of drugs. Flunixin meglumine and phenylbutazone have been shown to have a favorable influence on the course and outcome of certain diseases. This report reviews information concerning the pharmacology, pharmacokinetics, and therapeutics of phenylbutazone and flunixin as they have been used on an experimental basis in food animals.
Sigma-Aldrich
Flunixin meglumine, ≥98% (HPLC)
Flunixin meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenylbutazone
Supelco
Phenylbutazone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Phenylbutazone, European Pharmacopoeia (EP) Reference Standard