Merck
Fungal zinc metabolism and its connections to virulence.
Frontiers in cellular and infection microbiology (2013-10-18)
Charley C Staats, Lívia Kmetzsch, Augusto Schrank, Marilene H Vainstein
ABSTRACT
Zinc is a ubiquitous metal in all life forms, as it is a structural component of the almost 10% of eukaryotic proteins, which are called zinc-binding proteins. In zinc-limiting conditions such as those found during infection, pathogenic fungi activate the expression of several systems to enhance the uptake of zinc. These systems include ZIP transporters (solute carrier 39 family) and secreted zincophores, which are proteins that are able to chelate zinc. The expression of some fungal zinc uptake systems are regulated by a master regulator (Zap1), first characterized in the yeast Saccharomyces cerevisiae. In this review, we highlight features of zinc uptake and metabolism in human fungal pathogens and aspects of the relationship between proper zinc metabolism and the expression of virulence factors and adaptation to the host habitat.
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Zinc, wire reel, 10m, diameter 0.5mm, as drawn, 99.99+%
Zinc, wire reel, 5m, diameter 0.05mm, as drawn, 99%
Zinc, wire reel, 0.1m, diameter 6.35mm, 99.99+%
Zinc, wire reel, 1m, diameter 1.0mm, 99.99+%
Zinc, wire reel, 5m, diameter 0.025mm, as drawn, 99%
Zinc, wire reel, 100m, diameter 2.0mm, extruded, 99.9%
Zinc, wire reel, 25m, diameter 0.125mm, as drawn, 99%
Zinc, wire reel, 20m, diameter 2.0mm, extruded, 99.9%
Zinc, wire reel, 10m, diameter 0.25mm, as drawn, 99.99+%
Zinc, wire reel, 2m, diameter 1.0mm, 99.999%
Zinc, foil, 8mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.015mm, 99.9%
Zinc, foil, not light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 5m coil, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95+%