Merck
The governance dimensions of water security: a review.
Philosophical transactions. Series A, Mathematical, physical, and engineering sciences (2013-10-02)
Karen Bakker, Cynthia Morinville
ABSTRACT
Water governance is critical to water security, and to the long-term sustainability of the Earth's freshwater systems. This review examines recent debates regarding the governance dimensions of water security, including adaptive governance, polycentric governance, social learning and multi-level governance. The analysis emphasizes the political and institutional dimensions of water governance, and explores the relevance of social power-an overlooked yet important aspect of the water security debate. In addition, the review explores the intersection and potential synergies between water governance perspectives and risk-based approaches to water security, and offers critiques and suggestions for further research questions and agendas.
Product Description
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, suitable for HPLC
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, suitable for ion chromatography
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O