Merck
  • XRCC3 Thr241Met polymorphism and clinical outcomes of NSCLC patients receiving platinum-based chemotherapy: a systematic review and meta-analysis.
XRCC3 Thr241Met polymorphism and clinical outcomes of NSCLC patients receiving platinum-based chemotherapy: a systematic review and meta-analysis.
PloS one (2013-08-14)
Xiao-yong Shen, Fan-zhen Lu, Yun Wu, Li-ting Zhao, Zhi-feng Lin
ABSTRACT
X-ray repair cross-complementing protein 3 (XRCC3) is an essential gene involved in the double-strand break repair pathway. Published evidence has shown controversial results about the relationship between XRCC3 Thr241Met polymorphism and clinical outcomes of non-small cell lung cancer (NSCLC) patients receiving platinum-based chemotherapy. A systematic review and meta-analysis was performed to evaluate the predictive value of XRCC3 Thr241Met polymorphism on clinical outcomes of advanced NSCLC receiving platinum-based chemotherapy. Response to chemotherapy, overall survival (OS) and progression-free survival (PFS) were analyzed. A number of 11 eligible studies were identified according to the inclusion criteria. Carriers of the variant XRCC3 241Met allele were significantly associated with good response to platinum-based chemotherapy (ThrMet/MetMet vs. ThrThr: OR  = 1.509, 95% CI: 1.099-2.072, Pheterogeneity  = 0.618). The XRCC3 Thr241Met polymorphism was not associated with OS (MetMet vs. ThrThr, HR  = 0.939, 95% CI:0.651-1.356, Pheterogeneity  = 0.112) or PFS (MetMet vs. ThrThr, HR  = 0.960, 95% CI: 0.539-1.710, Pheterogeneity  = 0.198). Additionally, no evidence of publication bias was observed. This systematic review and meta-analysis shows that carriers of the XRCC3 241Met allele are associated with good response to platinum-based chemotherapy in advanced NSCLC, while the XRCC3 Thr241Met polymorphism is not associated with OS or PFS.
Sigma-Aldrich
Platinum, nanopowder, <50 nm particle size (TEM)
Platinum, foil, 0.1m coil, thickness 0.025mm, coil width 8mm, as rolled, 99.95%
Sigma-Aldrich
Platinum, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.10 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Platinum, foil, 25x25mm, thickness 0.25mm, as rolled, 99.999%
Platinum, wire reel, 2m, diameter 0.25mm, annealed, may be used as one component of a thermocouple, 99.99%
Platinum, insulated wire, 0.5m, conductor diameter 0.125mm, insulation thickness 0.016mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Platinum, wire reel, 0.1m, diameter 0.005mm, wollaston, 99.9%
Platinum, insulated wire, 1m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%
Platinum, tube, 100mm, outside diameter 1.2mm, inside diameter 1.0mm, wall thickness 0.1mm, as drawn, 99.95%
Platinum, wire reel, 0.2m, diameter 0.8mm, annealed, may be used as one component of a thermocouple, 99.99%
Platinum, wire reel, 2m, diameter 0.20mm, annealed, may be used as one component of a thermocouple, 99.99%
Platinum, rod, 6.5mm, diameter 6.0mm, 99.99+%
Platinum, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95%
Platinum, insulated wire, 0.5m, conductor diameter 0.018mm, insulation thickness 0.002mm, polyester insulation, 99.99%
Platinum, wire reel, 10m, diameter 0.02mm, as drawn, 99.9%
Platinum, wire reel, 0.1m, diameter 0.40mm, as drawn, 99.99%
Platinum, foil, 0.1m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Platinum, wire reel, 0.1m, diameter 0.001mm, wollaston, 99.9%
Platinum, microfoil, disks, 25mm, thinness 1.0μm, specific density 900μg/cm2, permanent mylar 3.5μm support, 99.99+%
Platinum, foil, not light tested, 100x100mm, thickness 0.0075mm, as rolled, 99.95%
Sigma-Aldrich
Platinum, powder, 99.995% trace metals basis
Sigma-Aldrich
Platinum, powder (coarse), 99.99% trace metals basis
Sigma-Aldrich
Platinum, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis