- Silver as antibacterial agent: ion, nanoparticle, and metal.
Silver as antibacterial agent: ion, nanoparticle, and metal.
Angewandte Chemie (International ed. in English) (2012-12-21)
Svitlana Chernousova, Matthias Epple
PMID23255416
ABSTRACT
The antibacterial action of silver is utilized in numerous consumer products and medical devices. Metallic silver, silver salts, and also silver nanoparticles are used for this purpose. The state of research on the effect of silver on bacteria, cells, and higher organisms is summarized. It can be concluded that the therapeutic window for silver is narrower than often assumed. However, the risks for humans and the environment are probably limited.
Product Description
Silver, tube, 100mm, outside diameter 1.1mm, inside diameter 0.75mm, wall thickness 0.175mm, as drawn, 99.95+%
Silver, rod, 50mm, diameter 16mm, as drawn, 99.95+%
Silver, rod, 100mm, diameter 2.0mm, as drawn, 99.95+%
Silver, tube, 200mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.95+%
Silver, rod, 100mm, diameter 3.5mm, as drawn, 99.95+%
Silver, tube, 200mm, outside diameter 10mm, inside diameter 8mm, wall thickness 1.0mm, as drawn, 99.95+%
Silver, sphere, 100pcs, diameter 0.5mm, precision sphere grade
Silver, tube, 200mm, outside diameter 4.65mm, inside diameter 4.15mm, wall thickness 0.25mm, as drawn, 99.95+%
Silver, rod, 50mm, diameter 6.0mm, as drawn, 99.95+%
Silver, rod, 200mm, diameter 6.0mm, as drawn, 99.95+%
Silver, rod, 1000mm, diameter 5.0mm, as drawn, 99.95+%
Silver, foil, not light tested, 300x300mm, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%
Silver, foil, light tested, 50x50mm, thickness 0.0125mm, 99.95+%
Silver, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 50mm disks, thickness 0.125mm, annealed, 99.95+%
Silver, foil, 4mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 4mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 25mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 4mm disks, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 4mm disks, thickness 0.007mm, 99.97%