Merck
  • Urgent liver transplantation for nevirapine-induced acute liver failure: report of a case and review of the literature.
Urgent liver transplantation for nevirapine-induced acute liver failure: report of a case and review of the literature.
Annals of transplantation (2012-04-03)
Juan Carlos Lopez-Delgado, Rafael Mañez Mendiluce, Teresa Serrano Pinol, Xose Pérez Fernández, Laura Sanchez, Rosa Granada Vicente
ABSTRACT
Acute liver failure (ALF) due to antiretroviral treatment is a rare but a fatal complication in HIV-infected patients. Non-nucleoside retroviral transcriptase inhibitors are known to cause hepatotoxicity and exceptionally acute hepatic failure. We report a case of nevirapine-induced acute liver failure successfully managed with orthotopic liver transplantation (OLT). Antiretroviral therapy including raltegravir, zidovudine and abacavir was reintroduced 6 days after transplantation. No interaction with immunosuppressive agents was observed and the patient had a complete recovery. Urgent OLT should be considered in HIV patients with nevirapine-induced ALF because of the poor outcome despite intensive supportive conservative treatment. Concomitant use of highly active antiretroviral therapy (HAART) and immunosuppression could be feasible if treatment is performed cautiously.
Product Description
Nevirapine for peak identification, European Pharmacopoeia (EP) Reference Standard
Nevirapine (anhydrous), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nevirapine