Merck
  • Review article: rifabutin in the treatment of refractory Helicobacter pylori infection.
Review article: rifabutin in the treatment of refractory Helicobacter pylori infection.
Alimentary pharmacology & therapeutics (2011-12-02)
J P Gisbert, X Calvet
ABSTRACT
Even with the current most effective treatment regimens, a relevant proportion of patients will fail to eradicate Helicobacter pylori infection. To evaluate the role of rifabutin in the treatment of H. pylori infection. Bibliographical searches were performed in MEDLINE. Data on the efficacy of rifabutin-containing regimens on H. pylori eradication were combined and meta-analysed using the generic inverse variance method. Rifabutin shows good in vitro activity against H. pylori. Mean H. pylori rifabutin resistance rate (calculated from 11 studies including 2982 patients) was 1.3% (95% confidence interval = 0.9-1.7%). When only studies including patients naïve to H. pylori eradication treatment were considered, this figure was even lower (0.6%). On the other hand, higher values of rifabutin resistance were calculated (1.59%) when only post-treatment patients were considered. Overall, mean H. pylori eradication rate (intention-to-treat analysis) with rifabutin-containing regimens (1008 patients) was 73% (67-79%). Respective cure rates for second-line (223 patients), third-line (342 patients) and fourth/fifth-line (95 patients) rifabutin therapies were 79% (67-92%), 66% (55-77%) and 70% (60-79%) respectively. For treating H. pylori infection, almost all studies have administered rifabutin 300 mg/day; this dose seems to be more effective than 150 mg/day. The ideal length of treatment remains unclear, but 10- to 12-day regimens are generally recommended. The mean rate of adverse effects was 22% (19-25%). Myelotoxicity is the most significant, although this complication was rare. Until now, all patients have recovered of leucopenia uneventfully in a few days, and there have been no reports of infection or other adverse outcomes related to it. Rifabutin-containing rescue therapy constitutes an encouraging strategy after multiple (usually three) previous eradication failures with key antibiotics such as amoxicillin, clarithromycin, metronidazole, tetracycline and levofloxacin.
Sigma-Aldrich
Rifabutin, >98% (HPLC), powder
Pricing and availability is not currently available.
USP
Rifabutin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Rifabutin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.