Merck
Palladium-catalysed hydroxylation and alkoxylation.
Chemical Society reviews (2011-06-07)
Stephan Enthaler, Anna Company
ABSTRACT
The formation of oxygen-carbon bonds is one of the fundamental transformations in organic synthesis. In this regard the application of palladium-based catalysts has been extensively studied during recent years. Nowadays it is an established methodology and the success has been proven in manifold synthetic procedures. This tutorial review summarizes the advances on palladium-catalysed C-O bond formation, means hydroxylation and alkoxylation reactions.
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, 25x25mm, thickness 0.50mm, as rolled, 99.99+%
Palladium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Palladium, foil, 4mm disks, thickness 0.02mm, as rolled, 99.95%
Palladium, foil, 15mm disks, thickness 0.075mm, as rolled, 99.95%
Palladium, foil, 10mm disks, thickness 0.0125mm, 99.95%
Palladium, foil, 10x10mm, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, 6mm disks, thickness 0.0125mm, 99.95%
Palladium, foil, 4mm disks, thickness 0.008mm, 99.95%
Palladium, foil, 10mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%
Palladium, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, 10x10mm, thickness 0.50mm, as rolled, 99.99+%