Merck
Contact allergy to furazolidone.
Contact dermatitis (1990-04-01)
A C de Groot, J M Conemans
ABSTRACT
A case of occupational contact allergy to furazolidone, used as an animal feed additive and as an antimicrobial drug in veterinary medicine, is described. The patient did not react to furazolidone 2% pet. Using PEG-400 and alcohol as patch test vehicles resulted in positive patch test reactions. No cross-reactions were observed to other nitrofuran derivatives (nitrofurazone, nitrofurantoin) or to furfural. The literature on contact allergy to nitrofurans is reviewed.
Sigma-Aldrich
Furazolidone